Learn More
Aviva Systems Biology Corp CCL18 Antibody - middle region (AVARP07049_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP07049_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CCL18. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human CCL18. Alternate names: CKb7, PARC, AMAC1, DCCK1, MIP-4, AMAC-1, DC-CK1, SCYA18ApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-53775