Learn More
American Radiolabeled Chemicals Inc CCL2-125I 25UC
SDP
Catalog No. 50901047
Encompass
MCP-1-125I Bolton-Hunter Labeled 25uCi in buffered solution, 0.025 mCi/ml, S.A. 2200 Ci/mmol, M.W. ~8600, Shipped in Dry Ice
Catalog No. 50-901-047 Supplier American Radiolabeled Chemicals Inc Supplier No. ARI 015525UC
Add to Cart
Add to Cart
Request a Quote
MCP-1-125I Bolton-Hunter Labeled 25uCi in buffered solution, 0.025 mCi/ml, S.A. 2200 Ci/mmol, M.W. ~8600, Shipped in Dry Ice