Learn More
American Radiolabeled Chemicals Inc CCL2-125I 5UC
SDP
Catalog No. 50901048
Encompass
MCP-1-125I Bolton-Hunter Labeled 5uCi in buffered solution, 0.025 mCi/ml, S.A. 2200 Ci/mmol, M.W. ~8600, Shipped in Dry Ice
Catalog No. 50-901-048 Supplier American Radiolabeled Chemicals Inc Supplier No. ARI 01555UC
MCP-1-125I Bolton-Hunter Labeled 5uCi in buffered solution, 0.025 mCi/ml, S.A. 2200 Ci/mmol, M.W. ~8600, Shipped in Dry Ice