Learn More
Aviva Systems Biology Corp CCL2 Antibody - middle region (AVARP07046_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP07046_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CCL2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CCL2. Alternate names: GDCF-2, GDCF-2 HC11, HC11, HSMCR30, MCAF, MCP-1, MCP1, MGC9434, SCYA2, SMC-CFApplicationIHC, WB. Species reactivity: Dog, Human
Catalog No. 50-014-62553