Learn More
Enzo Life Sciences CCL2 (human) monoclonal antibody (5D3-F7) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804465C100
Encompass_Preferred
Clone: 5D3-F7. Isotype: IgG1. Immunogen: Recombinant human CCL2 (monocyte chemotactic protein-1; MCP-1). Species reactivity: Human. Applications: ELISA, ICC, IHC (FS), IHC (PS), IP, WB. Formulation: Liquid. In PBS containing 0.1% sodium azide.
Catalog No. 50-200-9917