Learn More
Sino Biological CCL2 / MCP-1 Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 50368T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CCL2 / MCP-1 (rM CCL2 / MCP-1; Catalog#50368-M08B; NP_035463.1; Met1-Asn148). CCL2 / MCP-1 specific IgG was purified by Mouse CCL2 / MCP-1 affinity chromatography.
Catalog No. 50-224-7540