Learn More
Enzo Life Sciences CCL22 (human) monoclonal antibody (349/6) (250µg)
SDP
Supplier:  Enzo Life Sciences ALX804469C250
Encompass_Preferred
Clone: 349/6. Isotype: IgG. Immunogen: Recombinant human CCL22 (macrophage-derived chemokine; MDC). Species reactivity: Human. Applications: ELISA, WB. Purity: Protein G-affinity purified. Formulation: Liquid. 0.2μm-filtered solution in PBS, pH 7.4. Contains no preservatives.
Catalog No. 50-200-9918