Learn More
Medchemexpress LLC CCL4 Human 100ug
SDP
Catalog No. 5000182845
Encompass_Preferred
CCL4 Protein Human is a small cytokine of the CC chemokine subfamily that binds to the CCR5 chemokine receptor on the cell surface promotes leukocyte aggregation under various inflammatory conditions and contributes to immune protection against human immunodeficiency virus type 1 CCL4 Protein Human is a recombinant human CCL4 (A24-N92) expressed by E coli[1]
Catalog No. 50-001-82845 Supplier Medchemexpress LLC Supplier No. HYP7257100UG
CCL4 Protein Human is a small cytokine of the CC chemokine subfamily that binds to the CCR5 chemokine receptor on the cell surface promotes leukocyte aggregation under various inflammatory conditions and contributes to immune protection against human immunodeficiency virus type 1 CCL4 Protein Human is a recombinant human CCL4 (A24-N92) expressed by E coli[1]