Learn More
Medchemexpress LLC CCL4 Human 10ug
SDP
Catalog No. 5000181490
Encompass_Preferred
CCL4 Protein Human is a small cytokine of the CC chemokine subfamily that binds to the CCR5 chemokine receptor on the cell surface promotes leukocyte aggregation under various inflammatory conditions and contributes to immune protection against human immunodeficiency virus type 1 CCL4 Protein Human is a recombinant human CCL4 (A24-N92) expressed by E coli[1]
Catalog No. 50-001-81490 Supplier Medchemexpress LLC Supplier No. HYP725710UG
CCL4 Protein Human is a small cytokine of the CC chemokine subfamily that binds to the CCR5 chemokine receptor on the cell surface promotes leukocyte aggregation under various inflammatory conditions and contributes to immune protection against human immunodeficiency virus type 1 CCL4 Protein Human is a recombinant human CCL4 (A24-N92) expressed by E coli[1]