Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CCL4 Mouse 10ug
SDP

Catalog No. 5000196220
Encompass_Preferred

Recombinant mouse CCL4 (C-C motif chemokine ligand 4) is a proinflammatory chemokine expressed in E. coli and supplied lyophilized for research use. It promotes leukocyte chemotaxis, acts as a monokine that can form homodimers, and is provided with high purity suitable for in vitro studies of inflammation and immune cell migration.

  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Recombinant expression in E. coli without affinity tag.
  • Supplied lyophilized from buffered formulation for stability.
  • Predicted molecular mass ~7.8 kDa (observed ~5-10 kDa by SDS-PAGE).
  • Intended for in vitro research applications studying chemotaxis and inflammation.
  • Stable when aliquoted and stored at -20°C or -80°C for extended storage.

Catalog No. 50-001-96220 Supplier Medchemexpress LLC Supplier No. HYP7188910UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse CCL4 (C-C motif chemokine ligand 4) is a proinflammatory chemokine expressed in E. coli and supplied lyophilized for research use. It promotes leukocyte chemotaxis, acts as a monokine that can form homodimers, and is provided with high purity suitable for in vitro studies of inflammation and immune cell migration.

  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Recombinant expression in E. coli without affinity tag.
  • Supplied lyophilized from buffered formulation for stability.
  • Predicted molecular mass ~7.8 kDa (observed ~5-10 kDa by SDS-PAGE).
  • Intended for in vitro research applications studying chemotaxis and inflammation.
  • Stable when aliquoted and stored at -20°C or -80°C for extended storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.