Learn More
Cayman Chemical CCL5RnTEShumnrecombIna 100ug
SDP
Catalog No. 502950475
Encompass_Preferred
Source: Recombinant human CCL5 expressed in E. coli • Amino acids: 24-91 • MW: 8 kDa
Catalog No. 50-295-0475 Supplier Cayman Chemical Supplier No. 32858100 UG
Source: Recombinant human CCL5 expressed in E. coli • Amino acids: 24-91 • MW: 8 kDa