Learn More
Medchemexpress LLC C-C motif chemokine 6 (rat), N-terminal His-tag | 100UG
SDP
Catalog No. 5000176285
Encompass_Preferred
Recombinant rat CCL6 chemokine with an N-terminal His tag, expressed in E. coli and supplied as a lyophilized powder for biochemical and cell-based research. The protein shows documented biological activity, low endotoxin, and is formulated for stability and storage at -20°C.
Catalog No. 50-001-76285 Supplier Medchemexpress LLC Supplier No. HYP700281100UG
Recombinant rat CCL6 chemokine with an N-terminal His tag, expressed in E. coli and supplied as a lyophilized powder for biochemical and cell-based research. The protein shows documented biological activity, low endotoxin, and is formulated for stability and storage at -20°C.