Learn More
Medchemexpress LLC C-C motif chemokine 6 (Ccl6) | 10UG
SDP
Catalog No. 5000176286
Encompass_Preferred
Recombinant rat CCL6 protein with a histidine affinity tag, expressed in E. coli and supplied lyophilized for research use. The protein corresponds to residues G22-A115 of accession Q68FP3 and demonstrates biological activity in endothelial cell assays.
  • His-tagged recombinant rat CCL6, residues G22-A115.
  • Expressed in E. coli and lyophilized from 50 mM Tris-HCl, 300 mM NaCl, pH 7.4.
  • Approximate molecular weight ~12 kDa by SDS-PAGE.
  • Bioactive in HUVEC proliferation assay; ED50 15.84 ng/mL.
  • Stable when stored at -20°C; aliquot and freeze at -20°C or -80°C for long-term storage.
  • Supplied in a 10 μg vial for small-scale experiments.
Catalog No. 50-001-76286 Supplier Medchemexpress LLC Supplier No. HYP70028110UG
Recombinant rat CCL6 protein with a histidine affinity tag, expressed in E. coli and supplied lyophilized for research use. The protein corresponds to residues G22-A115 of accession Q68FP3 and demonstrates biological activity in endothelial cell assays.
  • His-tagged recombinant rat CCL6, residues G22-A115.
  • Expressed in E. coli and lyophilized from 50 mM Tris-HCl, 300 mM NaCl, pH 7.4.
  • Approximate molecular weight ~12 kDa by SDS-PAGE.
  • Bioactive in HUVEC proliferation assay; ED50 15.84 ng/mL.
  • Stable when stored at -20°C; aliquot and freeze at -20°C or -80°C for long-term storage.
  • Supplied in a 10 μg vial for small-scale experiments.