Learn More
Medchemexpress LLC CCL6 Rat N-His 2ug
SDP
Catalog No. 5000177264
Encompass_Preferred
The CCL6 protein is part of the intercrine beta family a component of chemokines involved in intercellular communication and immune responses In this family CCL6 may play an important role in regulating inflammatory processes and cellular interactions CCL6 Protein Rat (N-His) is the recombinant rat-derived CCL6 protein expressed by E coli with N-6 His labeled tag
Catalog No. 50-001-77264 Supplier Medchemexpress LLC Supplier No. HYP7002812UG
The CCL6 protein is part of the intercrine beta family a component of chemokines involved in intercellular communication and immune responses In this family CCL6 may play an important role in regulating inflammatory processes and cellular interactions CCL6 Protein Rat (N-His) is the recombinant rat-derived CCL6 protein expressed by E coli with N-6 His labeled tag