Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC C-C motif chemokine 6 (rat), recombinant, N-terminal His tag | >95.0% | 50UG
SDP

Catalog No. 5000177279
Encompass_Preferred

Recombinant rat C-C motif chemokine 6 (CCL6) with an N-terminal His tag, expressed in E. coli and provided as a lyophilized protein for immunology and cell-based assays. The product corresponds to residues G22-A115 of accession Q68FP3 and shows biological activity (ED50 15.84 ng/mL).

  • N-terminal His-tagged recombinant rat chemokine.
  • Amino acid range G22-A115 (accession Q68FP3).
  • Expression system: E. coli; lyophilized formulation from 50 mM Tris-HCl, 300 mM NaCl, pH 7.4.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Endotoxin level <1 EU/μg (LAL method).
  • Approximate molecular weight ~12 kDa.
  • Stable when stored at -20°C; reconstituted stability 4°C for 1 week or -20°C longer with carrier protein.

Catalog No. 50-001-77279 Supplier Medchemexpress LLC Supplier No. HYP70028150UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant rat C-C motif chemokine 6 (CCL6) with an N-terminal His tag, expressed in E. coli and provided as a lyophilized protein for immunology and cell-based assays. The product corresponds to residues G22-A115 of accession Q68FP3 and shows biological activity (ED50 15.84 ng/mL).

  • N-terminal His-tagged recombinant rat chemokine.
  • Amino acid range G22-A115 (accession Q68FP3).
  • Expression system: E. coli; lyophilized formulation from 50 mM Tris-HCl, 300 mM NaCl, pH 7.4.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Endotoxin level <1 EU/μg (LAL method).
  • Approximate molecular weight ~12 kDa.
  • Stable when stored at -20°C; reconstituted stability 4°C for 1 week or -20°C longer with carrier protein.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.