Learn More
Teknova CCMB80 Buffer. 500mL, Sterile.
SDP
Catalog No. NC1000864
Encompass
80mM Calcium Chloride,20mM Manganese Chloride,10mM Magnesium Chloride,10mM Potassium Acetate,10% Glycerol
Catalog No. NC1000864 Supplier Teknova Supplier No. C3132
80mM Calcium Chloride,20mM Manganese Chloride,10mM Magnesium Chloride,10mM Potassium Acetate,10% Glycerol