Learn More
Medchemexpress LLC HY-12150 5mg Medchemexpress, CCMI CAS:917837-54-8 Purity:>98%
SDP
Catalog No. 501932559
Encompass_Preferred
Medchemexpress, HY-12150 5mg CCMI CAS:917837-54-8 CCMI is a potent and selective α7 nAChR -positive allosteric modulator, does not bind to or activate α7 nAChRs via the orthosteric site, and causes significant positive modulation of agonist-induced currents at α7 nAChRs. CCMI has potential in CNS diseases with cognitive dysfunction [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2559 Supplier Medchemexpress LLC Supplier No. HY121505MG
Medchemexpress, HY-12150 5mg CCMI CAS:917837-54-8 CCMI is a potent and selective α7 nAChR -positive allosteric modulator, does not bind to or activate α7 nAChRs via the orthosteric site, and causes significant positive modulation of agonist-induced currents at α7 nAChRs. CCMI has potential in CNS diseases with cognitive dysfunction [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.