Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ccmi (synonyms: AVL-3288; UCI-4083) | 917837-54-8 | 100.0% | 250 MG
SDP

Catalog No. 5000231231
Encompass_Preferred

CCMI (AVL-3288) is a selective positive allosteric modulator of the α7 nicotinic acetylcholine receptor used in neuroscience research to potentiate agonist-induced currents. Supplied as a light yellow solid with high purity, it is provided in multiple pack sizes for laboratory use.

  • Potent α7 nAChR positive allosteric modulator.
  • Used in CNS and cognitive dysfunction research.
  • High purity: 99.97%.
  • Solid, light yellow to yellow appearance.
  • Molecular formula C19H15Cl2N3O2; molecular weight 388.25 g·mol⁻1.
  • Available in pack sizes including 250 MG.
  • Documentation available: data sheet, COA, and SDS.

Catalog No. 50-002-31231 Supplier Medchemexpress LLC Supplier No. HY12150250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CCMI (AVL-3288) is a selective positive allosteric modulator of the α7 nicotinic acetylcholine receptor used in neuroscience research to potentiate agonist-induced currents. Supplied as a light yellow solid with high purity, it is provided in multiple pack sizes for laboratory use.

  • Potent α7 nAChR positive allosteric modulator.
  • Used in CNS and cognitive dysfunction research.
  • High purity: 99.97%.
  • Solid, light yellow to yellow appearance.
  • Molecular formula C19H15Cl2N3O2; molecular weight 388.25 g·mol⁻1.
  • Available in pack sizes including 250 MG.
  • Documentation available: data sheet, COA, and SDS.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.