Learn More
Medchemexpress LLC CCN2/CTGF Human Bi 20ug | 20ug
SDP
Catalog No. 5000187525
Encompass_Preferred
CCN2/CTGF (Connective Tissue Growth Factor) is a protein produced primarily by endothelial cells in blood vessels It plays an important role in various cellular processes CCN2/CTGF Protein Human (Biotinylated HEK293 His-Avi) is the recombinant human-derived CCN2/CTGF protein expressed by HEK293 with C-Avi C-His labeled tag
Catalog No. 50-001-87525 Supplier Medchemexpress LLC Supplier No. HYP7810420UG
CCN2/CTGF (Connective Tissue Growth Factor) is a protein produced primarily by endothelial cells in blood vessels It plays an important role in various cellular processes CCN2/CTGF Protein Human (Biotinylated HEK293 His-Avi) is the recombinant human-derived CCN2/CTGF protein expressed by HEK293 with C-Avi C-His labeled tag