Learn More
Medchemexpress LLC CCN2/CTGF protein, human (HEK293, His) | >95.0% | 100UG
SDP
Catalog No. 5000175367
Encompass_Preferred
Recombinant human CCN2/CTGF protein (Q27-A349), expressed in HEK293 cells and supplied as a C-terminal His-tagged, lyophilized powder. The material is formulated from 0.22 μm filtered PBS with trehalose protectant, exhibits >95.0% purity by reducing SDS-PAGE, and shows an apparent molecular weight of 45-60 kDa due to glycosylation. It is intended for research use in biochemical assays and antibody binding studies.
  • Recombinant human protein expressed in HEK293 cells.
  • C-terminal His tag for detection and purification.
  • >95.0% purity by reducing SDS-PAGE.
  • Apparent molecular weight 45-60 kDa due to glycosylation.
  • Lyophilized formulation with trehalose for stability.
  • Stable for 2 years at -20°C; aliquot and freeze at -20°C or -80°C for extended storage.
Catalog No. 50-001-75367 Supplier Medchemexpress LLC Supplier No. HYP700701100UG
Recombinant human CCN2/CTGF protein (Q27-A349), expressed in HEK293 cells and supplied as a C-terminal His-tagged, lyophilized powder. The material is formulated from 0.22 μm filtered PBS with trehalose protectant, exhibits >95.0% purity by reducing SDS-PAGE, and shows an apparent molecular weight of 45-60 kDa due to glycosylation. It is intended for research use in biochemical assays and antibody binding studies.
  • Recombinant human protein expressed in HEK293 cells.
  • C-terminal His tag for detection and purification.
  • >95.0% purity by reducing SDS-PAGE.
  • Apparent molecular weight 45-60 kDa due to glycosylation.
  • Lyophilized formulation with trehalose for stability.
  • Stable for 2 years at -20°C; aliquot and freeze at -20°C or -80°C for extended storage.