Learn More
Medchemexpress LLC CCN2/CTGF protein, human (HEK293, His) | >95.0% | 50UG
SDP
Catalog No. 5000175363
Encompass_Preferred
Recombinant human CCN2/CTGF protein produced in HEK293 cells, supplied as a C-terminal His-tagged, lyophilized preparation. The protein is glycosylated and migrates at an apparent 45-60 kDa by SDS-PAGE, and is provided with high purity for use in cell biology and extracellular matrix research. Store lyophilized at -20°C and follow supplier instructions for reconstitution and handling.
  • Expressed in HEK293 cells.
  • C-terminal His tag for detection and purification.
  • Greater than 95.0% purity by reducing SDS-PAGE.
  • Apparent molecular weight 45-60 kDa due to glycosylation.
  • Lyophilized formulation for stability and long-term storage.
  • Suitable for cell signaling and extracellular matrix assays.
Catalog No. 50-001-75363 Supplier Medchemexpress LLC Supplier No. HYP70070150UG
Recombinant human CCN2/CTGF protein produced in HEK293 cells, supplied as a C-terminal His-tagged, lyophilized preparation. The protein is glycosylated and migrates at an apparent 45-60 kDa by SDS-PAGE, and is provided with high purity for use in cell biology and extracellular matrix research. Store lyophilized at -20°C and follow supplier instructions for reconstitution and handling.
  • Expressed in HEK293 cells.
  • C-terminal His tag for detection and purification.
  • Greater than 95.0% purity by reducing SDS-PAGE.
  • Apparent molecular weight 45-60 kDa due to glycosylation.
  • Lyophilized formulation for stability and long-term storage.
  • Suitable for cell signaling and extracellular matrix assays.