Learn More
Cayman Chemical CCN3humnrecombInt 100ug
SDP
Catalog No. 502945492
Encompass_Preferred
Source: Active recombinant human C-terminal His-tagged CCN3 expressed in insect cells • Amino acids: 1-357 • MW: 37 kD
Catalog No. 50-294-5492 Supplier Cayman Chemical Supplier No. 41888100 UG
Source: Active recombinant human C-terminal His-tagged CCN3 expressed in insect cells • Amino acids: 1-357 • MW: 37 kD