Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 891MSM2P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: CCNB1/1098. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: Cell line in logarithmic growth phase. Tonsil or testicular, endometrial, prostate or ovarian carcinoma. Cellular Staining: Cytoplasmic & Nuclear. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human full-length CCNB1 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 55-62kDa.Chromosome: 5q13.2. Gene Symbol: CCNB1. Gene ID: 891. Ensemble: ENSG00000134057. Uniprot: NP_114172.
Catalog No. 50-188-5314