Learn More
Aviva Systems Biology Corp CCNB2 Antibody - N-terminal region (ARP63410_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63410_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CCNB2. It was validated on Western Blot. . Alternate names: HsT17299ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rabbit, Rat
Catalog No. 50-287-962