Learn More
Aviva Systems Biology Corp CCNB3 Antibody - C-terminal region (ARP39740_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39740_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CCNB3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human CCNB3. Alternate names: -ApplicationWB. Species reactivity: Human
Catalog No. 50-014-57161