Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 595MSM2P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, WB, IHC. Conjugate/Tag: Unconjugated. Clone: CCND1/809. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2a kappa. Positive Control: T47D, ZR75, BT474, SKBR3, MCF-7, MDA-MB-453, HT29, Ramos, Jurkat or A431 cells. Mantle cell lymphoma, breast or bladder carcinomas. Cellular Staining: Nuclear. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human CCND1 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 36kDa.Chromosome: 11q13.2. Gene Symbol: CCND1. Gene ID: 595. Ensemble: ENSG00000110092. Uniprot: NP_444284.
Catalog No. 50-188-5132