Learn More
Aviva Systems Biology Corp CCND1 Antibody - N-terminal region (ARP33370_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33370_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CCND1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CCND1. Alternate names: BCL1, D11S287E, PRAD1, U21B31ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54984