Learn More
Aviva Systems Biology Corp CCND1 Antibody - middle region (AVARP03037_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP03037_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CCND1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CCND1. Alternate names: BCL1, D11S287E, PRAD1, U21B31ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-62520