Learn More
Aviva Systems Biology Corp CCND3 Antibody - C-terminal region (AVARP03038_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP03038_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CCND3. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human CCND3. Alternate names: ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rat, Sheep
Catalog No. 50-014-62521