Learn More
Medchemexpress LLC CCND3 protein, human (biotinylated, MBP, His, Flag) | >85.0% | 20UG
SDP
Catalog No. 5000174168
Encompass_Preferred
Recombinant human CCND3 protein, biotinylated and fused to an N-terminal MBP with His and Flag tags, expressed in E. coli. The construct corresponds to CCND3 residues M1-L292 and is supplied in small research quantities (example: 20 μg). Characterization by reducing SDS-PAGE shows an apparent molecular weight of ~80 kDa and purity greater than 85%; consult the Certificate of Analysis for lot-specific data and storage instructions.
  • Biotinylated for affinity capture and detection.
  • N-terminal MBP fusion improves solubility.
  • His and Flag tags enable purification and detection.
  • Includes CCND3 residues M1-L292 for target-specific studies.
  • Characterized by SDS-PAGE with >85% purity.
  • Provided in small research-use quantities suitable for assay development.
Catalog No. 50-001-74168 Supplier Medchemexpress LLC Supplier No. HYP70295720UG
Recombinant human CCND3 protein, biotinylated and fused to an N-terminal MBP with His and Flag tags, expressed in E. coli. The construct corresponds to CCND3 residues M1-L292 and is supplied in small research quantities (example: 20 μg). Characterization by reducing SDS-PAGE shows an apparent molecular weight of ~80 kDa and purity greater than 85%; consult the Certificate of Analysis for lot-specific data and storage instructions.
  • Biotinylated for affinity capture and detection.
  • N-terminal MBP fusion improves solubility.
  • His and Flag tags enable purification and detection.
  • Includes CCND3 residues M1-L292 for target-specific studies.
  • Characterized by SDS-PAGE with >85% purity.
  • Provided in small research-use quantities suitable for assay development.