Learn More
Medchemexpress LLC Ccnd3 protein, human (biotinylated, MBP, His, Flag) | >85.0% | 50UG
SDP
Catalog No. 5000174188
Encompass_Preferred
Recombinant human cyclin D3 (CCND3) is supplied as a biotinylated, affinity-tagged protein spanning amino acids M1-L292. The product is expressed in E. coli and formulated for research applications including binding assays, pull-downs, and antibody validation; consult the SDS/COA for storage and handling information.
  • Biotinylated for streptavidin capture and sensitive detection.
  • N-terminal MBP tag to enhance solubility and facilitate purification.
  • C-terminal 6xHis and Flag tags for affinity purification and versatile detection.
  • Full-length human sequence (M1-L292) preserves native epitopes.
  • Expressed in E. coli for reproducible recombinant production.
  • Purity greater than 85% as determined by reducing SDS-PAGE.
  • Available in small research sizes suitable for assay development.
Catalog No. 50-001-74188 Supplier Medchemexpress LLC Supplier No. HYP70295750UG
Recombinant human cyclin D3 (CCND3) is supplied as a biotinylated, affinity-tagged protein spanning amino acids M1-L292. The product is expressed in E. coli and formulated for research applications including binding assays, pull-downs, and antibody validation; consult the SDS/COA for storage and handling information.
  • Biotinylated for streptavidin capture and sensitive detection.
  • N-terminal MBP tag to enhance solubility and facilitate purification.
  • C-terminal 6xHis and Flag tags for affinity purification and versatile detection.
  • Full-length human sequence (M1-L292) preserves native epitopes.
  • Expressed in E. coli for reproducible recombinant production.
  • Purity greater than 85% as determined by reducing SDS-PAGE.
  • Available in small research sizes suitable for assay development.