Learn More
Aviva Systems Biology Corp CCNDBP1 Antibody - middle region (ARP55415_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55415_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CCNDBP1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human CCNDBP1. Alternate names: DIP1, GCIP, HHMApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-034