Learn More
Aviva Systems Biology Corp CCNG2 Antibody - N-terminal region (AVARP03032_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP03032_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CCNG2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CCNG2. Alternate names: ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Rat
Catalog No. 50-014-53772