Learn More
Aviva Systems Biology Corp CCNL2 Antibody - N-terminal region (ARP50695_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50695_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CCNL2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CCNL2. Alternate names: ANIA-6B, DKFZp761A1210, DKFZp762O195, HCLA-ISO, HLA-ISO, PCEE, SB138, CCNMApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60413