Learn More
Aviva Systems Biology Corp CCNO Antibody - N-terminal region (ARP64648_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64648_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CCNO. It was validated on Western Blot. . Alternate names: CCNU, FLJ22422, UDG2, UNG2ApplicationWB. Species reactivity: Horse, Human, Mouse, Pig, Rabbit
Catalog No. 50-288-324