Learn More
Aviva Systems Biology Corp CCNY Antibody - middle region (ARP47922_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47922_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CCNY. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CCNY. Alternate names: C10orf9, CBCP1, CFP1, CCNXApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53532