Learn More
Medchemexpress LLC HY-P2171A 1mg Medchemexpress, CCP peptide (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP2171A1MG
Encompass_Preferred
Medchemexpress, HY-P2171A 1mg CCP peptide (TFA) CAS: CCP peptide TFA is a synthetic cyclic citrullinated peptide (CCP) and used as the substrate for detecting anti-CCP antibodies serologically. CCP peptide TFA functions as a target for autoantibodies with a very high specificity for rheumatoid arthritis (RA). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0550