Learn More
Aviva Systems Biology Corp CCPG1 Antibody - middle region (ARP46493_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46493_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CCPG1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CCPG1. Alternate names: CPR8, KIAA1254ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast
Catalog No. 50-014-53480