Learn More
Medchemexpress LLC HY-124759 5mg Medchemexpress, CCR1 antagonist 9 CAS:1220026-26-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1247595MG
Encompass_Preferred
Medchemexpress, HY-124759 5mg CCR1 antagonist 9 CAS:1220026-26-5 CCR1 antagonist 9 is a potent and selective CCR1 antagonist with an IC50 of 6.8 nM in calcium flux assay. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0008