Learn More
Eurofins Discoverx ChemiScreen CCR1 Chemokine Receptor Membrane Preparation, 1 vial. (HTS005M)
Catalog No. 502005507
Encompass
The ChemiScreen CCR1 Chemokine Receptor Membrane Preparation is derived from proprietary stable recombinant cells to ensure high levels of CCR1 surface expression. ChemiScreen membrane preps can be used to screen for ligand binding affinities using a competitive radio-labeled ligand approach. Increased binding affinities will lead to higher radioactive signal. Membrane preps contain 1 mL of 1 mg/mL frozen membranes derived from a stably modified clonal cell line which was chosen for optimal receptor expression. For detailed product information, visit www.discoverx.com/membranepreps.
Catalog No. 50-200-5507 Supplier Eurofins Discoverx Supplier No. HTS005M
The ChemiScreen CCR1 Chemokine Receptor Membrane Preparation is derived from proprietary stable recombinant cells to ensure high levels of CCR1 surface expression. ChemiScreen membrane preps can be used to screen for ligand binding affinities using a competitive radio-labeled ligand approach. Increased binding affinities will lead to higher radioactive signal. Membrane preps contain 1 mL of 1 mg/mL frozen membranes derived from a stably modified clonal cell line which was chosen for optimal receptor expression. For detailed product information, visit www.discoverx.com/membranepreps.