Learn More
Aviva Systems Biology Corp CCR10 Antibody - middle region (ARP63519_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63519_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CCR10. It was validated on Western Blot. . Alternate names: GPR2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-287-995