Learn More
Eurofins Discoverx PathHunter CCR2 Bioassay Kit, 2-Plate. (93-0192Y2-00061)
Catalog No. 501999911
Encompass
The PathHunter CCR2 Bioassay kit provides an easy-to-use cell based assay to measure drug potency and detect neutralizing antibodies. This bioassay assesses ligand (e.g. MCP-1/CCL2) based activation of CCR2 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged CCR2 and EA-tagged β-Arrestin-2. Activation of CCR2 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. Bioassay kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, control agonist, detection reagent, and assay plates. For detailed product information, User manual and COA, visit www.discoverx.com/bioassay.
Catalog No. 50-199-9911 Supplier Eurofins Discoverx Supplier No. 930192Y200061
The PathHunter CCR2 Bioassay kit provides an easy-to-use cell based assay to measure drug potency and detect neutralizing antibodies. This bioassay assesses ligand (e.g. MCP-1/CCL2) based activation of CCR2 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged CCR2 and EA-tagged β-Arrestin-2. Activation of CCR2 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. Bioassay kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, control agonist, detection reagent, and assay plates. For detailed product information, User manual and COA, visit www.discoverx.com/bioassay.