Learn More
Enzo Life Sciences CCR2 (human) polyclonal antibody (200µg)
SDP
Supplier:  Enzo Life Sciences ALX210821C200
Encompass_Preferred
Host: Goat. Immunogen: Synthetic peptide corresponding to aa 17-39 (G17EEVTTFFDYDYGAPCHKFDVKQ39) of the N-terminal extracellular domain of human CCR2 (C-C chemokine receptor 2). Species reactivity: Human. Applications: ICC, IHC (PS), WB. Purity: Epitope-affinity purified IgG. Formulation: Liquid. In PBS containing 1mg/ml BSA and 0.1% sodium azide. Handling: For maximum product recovery after thawing, centrifuge the vial before opening the cap. Avoid freeze/thaw cycles.
Catalog No. 50-200-8167