Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ccr2 protein, mouse (N-His, C-Myc) | 93.0% | 100UG
SDP

Catalog No. 5000175467
Encompass_Preferred

Recombinant CCR2 protein (mouse) representing amino acids M1-A55, expressed in Escherichia coli and supplied with an N-terminal 10xHis tag and a C-terminal c-Myc tag. The preparation has an apparent molecular weight of ~16 kDa and purity greater than 90% by reducing SDS-PAGE. It is intended for in vitro biochemical and binding studies involving CCR2 and its ligands. Store lyophilized or frozen, and aliquot to avoid freeze-thaw.

  • Recombinant fragment corresponding to CCR2 (mouse)
  • N-terminal 10xHis and C-terminal c-Myc tags for detection and purification
  • Expressed in Escherichia coli for high-yield production
  • Approximate molecular weight 16 kDa
  • Purity greater than 90% as determined by reducing SDS-PAGE
  • Suitable for biochemical and binding assays
  • Stable when stored at -20°C or -80°C; aliquot to avoid freeze-thaw

Catalog No. 50-001-75467 Supplier Medchemexpress LLC Supplier No. HYP700541100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant CCR2 protein (mouse) representing amino acids M1-A55, expressed in Escherichia coli and supplied with an N-terminal 10xHis tag and a C-terminal c-Myc tag. The preparation has an apparent molecular weight of ~16 kDa and purity greater than 90% by reducing SDS-PAGE. It is intended for in vitro biochemical and binding studies involving CCR2 and its ligands. Store lyophilized or frozen, and aliquot to avoid freeze-thaw.

  • Recombinant fragment corresponding to CCR2 (mouse)
  • N-terminal 10xHis and C-terminal c-Myc tags for detection and purification
  • Expressed in Escherichia coli for high-yield production
  • Approximate molecular weight 16 kDa
  • Purity greater than 90% as determined by reducing SDS-PAGE
  • Suitable for biochemical and binding assays
  • Stable when stored at -20°C or -80°C; aliquot to avoid freeze-thaw

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.