Learn More
Medchemexpress LLC CCR2 protein, mouse (N-His, C-Myc) | >90.0% | 20UG
SDP
Catalog No. 5000174962
Encompass_Preferred
Recombinant mouse CCR2 protein supplied lyophilized as a 16 kDa fragment for biochemical and binding studies. The protein is expressed in E. coli and contains N-terminal His and C-Myc tags to enable purification and detection. Supplied amount is 20 μg and purity exceeds 90% by reducing SDS-PAGE.
  • Expressed in E. coli with N-terminal His and C-Myc tags.
  • High purity (>90%) confirmed by reducing SDS-PAGE.
  • Supplied lyophilized with stabilizers for improved storage.
  • Suitable for binding assays and biochemical studies.
  • Store at -20°C; reconstituted material is stable at 4°C for one week.
Catalog No. 50-001-74962 Supplier Medchemexpress LLC Supplier No. HYP70054120UG
Recombinant mouse CCR2 protein supplied lyophilized as a 16 kDa fragment for biochemical and binding studies. The protein is expressed in E. coli and contains N-terminal His and C-Myc tags to enable purification and detection. Supplied amount is 20 μg and purity exceeds 90% by reducing SDS-PAGE.
  • Expressed in E. coli with N-terminal His and C-Myc tags.
  • High purity (>90%) confirmed by reducing SDS-PAGE.
  • Supplied lyophilized with stabilizers for improved storage.
  • Suitable for binding assays and biochemical studies.
  • Store at -20°C; reconstituted material is stable at 4°C for one week.