Learn More
Medchemexpress LLC HY-50081 5mg Medchemexpress, CCR2-RA-[R] CAS:512177-83-2 Purity:>98%
SDP
Catalog No. 502082246
Encompass_Preferred
Medchemexpress, HY-50081 5mg CCR2-RA-[R] CAS:512177-83-2 CCR2-RA-[R] is an allosteric antagonist of the C-C chemokine receptor type 2 (CCR2) with an IC50 of 103 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2246 Supplier Medchemexpress LLC Supplier No. HY500815MG
Medchemexpress, HY-50081 5mg CCR2-RA-[R] CAS:512177-83-2 CCR2-RA-[R] is an allosteric antagonist of the C-C chemokine receptor type 2 (CCR2) with an IC50 of 103 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.