Learn More
Eurofins Discoverx PathHunter CCR2 Total GPCR Internalization U2OS Cell Line, 2 vials. (93-0679C3)
Supplier:  Eurofins Discoverx 930679C3
Encompass
The PathHunter CCR2 Total GPCR internalization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This cell line uses EFC technology to measure ligand-induced internalization of CCR2 via detection of localization to the early endosome. The included cell line overexpresses ProLink-tagged CCR2 and an EA-tagged early endosome protein. Upon ligand-induced receptor activation, resulting internalization of the CCR2 receptor to the early endosome leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1089
Add to Cart
Request a Quote