Learn More
Medchemexpress LLC Ccr4-351 | 2174938-70-4 | MFCD32701932 | 99.0% | 500.42 g/mol | C24H27Cl2N7O | 5 MG
SDP
Catalog No. 5000262911
Encompass_Preferred
CCR4-351 is a small-molecule, orally active CCR4 antagonist used for preclinical research. It shows potent and selective activity in cellular assays and has demonstrated antitumor efficacy in animal models. For research use only; consult the supplier datasheet and safety documents for handling, storage, and dosing guidance.
  • Orally active and selective CCR4 antagonist.
  • Demonstrated antitumor activity in vivo.
  • Potent activity in cellular assays (low-nanomolar IC50 reported).
  • High chemical purity suitable for research applications.
  • Provided as a solid form for in vitro and in vivo studies.
  • Refer to the product datasheet and safety documents for handling and dosing.
Catalog No. 50-002-62911 Supplier Medchemexpress LLC Supplier No. HY1313495MG
CCR4-351 is a small-molecule, orally active CCR4 antagonist used for preclinical research. It shows potent and selective activity in cellular assays and has demonstrated antitumor efficacy in animal models. For research use only; consult the supplier datasheet and safety documents for handling, storage, and dosing guidance.
  • Orally active and selective CCR4 antagonist.
  • Demonstrated antitumor activity in vivo.
  • Potent activity in cellular assays (low-nanomolar IC50 reported).
  • High chemical purity suitable for research applications.
  • Provided as a solid form for in vitro and in vivo studies.
  • Refer to the product datasheet and safety documents for handling and dosing.