Learn More
Medchemexpress LLC CCR4-351 | 2174938-70-4 | MFCD32701932 | 99.0% | C24H27Cl2N7O | 10MG
SDP
Catalog No. 5000260889
Encompass_Preferred
CCR4-351 is an orally active, potent, and selective chemokine receptor 4 (CCR4) antagonist for research use. It features a novel piperidinyl-azetidine motif and exhibits low-nanomolar activity in functional assays (reported IC50 ≈ 22 nM in calcium flux and ≈ 50 nM in CTX). The compound has shown antitumor activity in preclinical studies and is supplied at high purity for experimental applications.
  • Potent CCR4 antagonist with low-nanomolar activity.
  • Selective activity in CCR4 functional assays.
  • Demonstrated antitumor activity in preclinical models.
  • Suitable for in vitro and in vivo CCR4 pathway research.
  • High purity (≈98.95%) for reliable experimental results.
Catalog No. 50-002-60889 Supplier Medchemexpress LLC Supplier No. HY13134910MG
CCR4-351 is an orally active, potent, and selective chemokine receptor 4 (CCR4) antagonist for research use. It features a novel piperidinyl-azetidine motif and exhibits low-nanomolar activity in functional assays (reported IC50 ≈ 22 nM in calcium flux and ≈ 50 nM in CTX). The compound has shown antitumor activity in preclinical studies and is supplied at high purity for experimental applications.
  • Potent CCR4 antagonist with low-nanomolar activity.
  • Selective activity in CCR4 functional assays.
  • Demonstrated antitumor activity in preclinical models.
  • Suitable for in vitro and in vivo CCR4 pathway research.
  • High purity (≈98.95%) for reliable experimental results.