Learn More
Medchemexpress LLC CCR4-351 hydrochloride | 2174938-71-5 | 98.4% | C24H28Cl3N7O | 10MG
SDP
Catalog No. 5000260958
Encompass_Preferred
CCR4-351 hydrochloride is an orally active, potent, and selective CCR4 antagonist supplied as a hydrochloride salt for research use. It is provided as a high-purity solid suitable for in vitro and in vivo pharmacology studies, with stability data and supporting documentation for handling and storage.
  • High purity (98.4%) suitable for preclinical research.
  • Orally active CCR4 antagonist with reported antitumor activity in vivo.
  • Available in multiple small pack sizes for dose-ranging studies.
  • White to off-white solid with defined storage recommendations for stability.
  • Data sheet and safety data sheet available to support experimental use and compliance.
Catalog No. 50-002-60958 Supplier Medchemexpress LLC Supplier No. HY131349A10MG
CCR4-351 hydrochloride is an orally active, potent, and selective CCR4 antagonist supplied as a hydrochloride salt for research use. It is provided as a high-purity solid suitable for in vitro and in vivo pharmacology studies, with stability data and supporting documentation for handling and storage.
  • High purity (98.4%) suitable for preclinical research.
  • Orally active CCR4 antagonist with reported antitumor activity in vivo.
  • Available in multiple small pack sizes for dose-ranging studies.
  • White to off-white solid with defined storage recommendations for stability.
  • Data sheet and safety data sheet available to support experimental use and compliance.